Trimethylphosphine ≥99%

Supplier: BeanTown Chemical

Synonyms: Methyl phosphine

Ampuled under argon

134115-1G 134115-25G 134115-5G
BT134115-1GEA 55.64 USD
BT134115-1G BT134115-25G BT134115-5G
Trimethylphosphine ≥99%
Trimethylphosphine

CAS: 594-09-2; EC No: 209-823-1; MDL No: MFCD00008510 UN No: UN2845; Haz Class: 4.2; Packing Group: I Liquid; Linear Formula: (CH3)3P; MW: 76.08 Boiling Point: 38-40°; Flash point: -30°C (-22°F) Density (g/mL): 0.738; Refractive Index: 1.428 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.428
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₃H₉P
MW: 76.08
Boiling Pt: 37.8
Melting Pt: -85
Density: 0.738 g/mL
Flash Pt: –30 °C (–22 °F)
MDL Number: MFCD00008510
CAS Number: 594-09-2
EINECS: 209-823-1
ADR: 4.2 I

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR