Cobalt ≥99.5% (trace metals basis), powder 1.6 µm
Supplier: BeanTown Chemical
134105-100G
134105-25G
134105-500G
BT134105-100GEA
88
USD
BT134105-100G
BT134105-25G
BT134105-500G
Cobalt ≥99.5% (trace metals basis), powder 1.6 µm
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GF8750000
UN No: UN3089; Haz Class: 4.1; Packing Group: II
Powder; Molecular Formula: Co; MW: 58.93
Melting Point: 1495°; Boiling Point: 2900°
Density (g/mL): 8.92
- RTECS: GF8750000
- Powder
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 Melting Pt: 1495 Density: 8.92 g/mL |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 UN: 3089 ADR: 4.1 II Merck Index: 12,02488 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...