Hydrochloric acid 5.0 N in water volumetric solution

Supplier: BeanTown Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

134060-1L 134060-20L 134060-4L
BT134060-1LEA 47.36 USD
BT134060-1L BT134060-20L BT134060-4L
Hydrochloric acid 5.0 N in water volumetric solution
Hydrochloric acid

CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324; RTECS: MW4025000 UN No: UN1789; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: HCl; MW: 36.46


  • RTECS: MW4025000
  • Liquid

Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR