2-Propanol ≥99.7% for gas chromatography
Supplier: BeanTown Chemical
Synonyms:
Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
134050-1L
134050-4L
134050-4X1L
BT134050-1LEA
49.48
USD
BT134050-1L
BT134050-4L
BT134050-4X1L
2-Propanol ≥99.7% for gas chromatography
2-Propanol
CAS: 67-63-0; EC No: 200-661-7; MDL No: MFCD00011674; RTECS: NT8050000
UN No: UN1219; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)2CHOH ; Molecular Formula: C3H8O; MW: 60.10
Melting Point: -90°; Boiling Point: 82°; Flash point: 12°C (54°F)
Density (g/mL): 0.785; Refractive Index: 1.377
- RTECS: NT8050000
- Refractive index: 1.377
- Liquid
Formula:
(CH₃)₂CHOH MW: 60.1 g/mol Boiling Pt: 82 Melting Pt: -89 Density: 0.785 g/mL Flash Pt: 12 °C (54 °F) |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3 II Merck Index: 13,05228 |
|
Specification Test Results
Assay | 99.7% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...