4-tert-Butylpyrocatechol ≥98%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol
133385-1KG
133385-250G
133385-5KG
BT133385-1KGEA
175.91
USD
BT133385-1KG
BT133385-250G
BT133385-5KG
4-tert-Butylpyrocatechol ≥98%, Technical Grade
4-tert-Butylpyrocatechol
CAS: 98-29-3; EC No: 202-653-9; MDL No: MFCD00002201; RTECS: UX1400000
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III
Flakes; Molecular Formula: C10H14O2; MW: 166.22
Melting Point: 53-56°; Boiling Point: 285°; Flash point:
- RTECS: UX1400000
- Flakes
Formula:
C₁₀H₁₄O₂ Boiling Pt: 285 °C Melting Pt: 53…56 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00002201 CAS Number: 98-29-3 EINECS: 202-653-9 UN: 2923 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...