Aluminum oxalate hydrate ≥99%

Supplier: BeanTown Chemical

133095-10G 133095-250G 133095-50G
BT133095-10GEA 27 USD
BT133095-10G BT133095-250G BT133095-50G
Aluminum oxalate hydrate ≥99%
Aluminum oxalate hydrate

CAS: 814-87-9; EC No: 212-407-2; MDL No: MFCD00050791; RTECS: BD0516250 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder/Chunks; Molecular Formula: C6Al2O12·xH2O; MW: 318.02 (anhydrous)


  • RTECS: BD0516250
  • Powder/Chunks

Formula: C₆Al₂O₁₂·xH₂O
MW: 318.017 g/mol (anhy)
MDL Number: MFCD00050791
CAS Number: 814-87-9
EINECS: 212-407-2
ADR: 6.1 III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR