Potassium hydroxide 0.1 N in methanol volumetric solution
Supplier: BeanTown Chemical
Synonyms:
Potash caustic, Potassium hydrate, Caustic potash
132705-1L
132705-20L
132705-4L
BT132705-1LEA
55.32
USD
BT132705-1L
BT132705-20L
BT132705-4L
Potassium hydroxide 0.1 N in methanol volumetric solution
Potassium hydroxide
CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: II
Liquid; Linear Formula: KOH; MW: 56.11
Flash point: 11°C (52°F)
Density (g/mL): 0.81
- Liquid
Formula:
KOH MW: 56.11 g/mol Boiling Pt: 1320 Melting Pt: 360 Density: 0.81 g/mL Flash Pt: 11 °C (52 °F) |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1992 ADR: 3 II Merck Index: 12,07806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...