Hydrochloric acid 36% (w/w) in water
Supplier: BeanTown Chemical
132495-1L
132495-2.5L
BT132495-1LEA
39.65
USD
BT132495-1L
BT132495-2.5L
Hydrochloric acid 36% (w/w) in water
Hydrochloric acid
CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324; RTECS: MW4025000
UN No: UN1789; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: HCl; MW: 36.46
Boiling Point: 81.5-110°
Density (g/mL): 1.19
- RTECS: MW4025000
- Liquid
Formula:
HCl Boiling Pt: 81.5…110 °C Density: 1.19 g/mL |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...