Molybdenum pentachloride ≥99.5%

Supplier: BeanTown Chemical

132360-250G 132360-50G 132360-5G
BT132360-250GEA 313 USD
BT132360-250G BT132360-50G BT132360-5G
Molybdenum pentachloride ≥99.5%
Molybdenum pentachloride

CAS: 10241-05-1; EC No: 233-575-3; MDL No: MFCD00003467; RTECS: QA4690000 UN No: UN2508; Haz Class: 8; Packing Group: III Crystalline/Powder; Linear Formula: MoCl5; MW: 273.21 Melting Point: 194°; Boiling Point: 268° Density (g/mL): 2.928 Air Sensitive, Moisture Sensitive


  • RTECS: QA4690000
  • Air Sensitive, Moisture Sensitive
  • Crystalline/Powder

Formula: Cl₅Mo
MW: 273.2 g/mol
Boiling Pt: 268
Melting Pt: 194
Density: 2.928 g/mL
MDL Number: MFCD00003467
CAS Number: 10241-05-1
EINECS: 233-575-3
UN: 2508
ADR: 8 III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR