Carbon black 50% in acetylene , 99.9%, compressed
Supplier: BeanTown Chemical
132335-1KG
132335-250G
132335-3KG
BT132335-1KGEA
197.94
USD
BT132335-1KG
BT132335-250G
BT132335-3KG
Carbon black 50% in acetylene , 99.9%, compressed
Carbon black
CAS: 1333-86-4; EC No: 215-609-9; MDL No: MFCD00133992; RTECS: FF5800000
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Solid, S.A. 75 m2/g, Bulk density 80-120g/L; Molecular Formula: C; MW: 12.01
Melting Point: 3550°
- RTECS: FF5800000
- Solid, S.A. 75 m²/g, Bulk density 80-120g/L
Formula:
C Melting Pt: 3550 °C Density: 80…120 g/L |
MDL Number:
MFCD00133992 CAS Number: 1333-86-4 EINECS: 215-609-9 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...