Silver hexafluoroantimonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Silver hexafluoroantimonate(V)

132075-1G 132075-25G 132075-5G
BT132075-1GEA 21 USD
BT132075-1G BT132075-25G BT132075-5G
Silver hexafluoroantimonate ≥99%
Silver hexafluoroantimonate

CAS: 26042-64-8; EC No: 247-429-1; MDL No: MFCD00003401 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: AgSbF6; MW: 343.61 Hygroscopic , Light Sensitive


  • Hygroscopic, Light Sensitive
  • Powder

Formula: AgF₆Sb
MDL Number: MFCD00003401
CAS Number: 26042-64-8
EINECS: 247-429-1
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR