Potassium hydroxide 30% w/v water

Supplier: BeanTown Chemical

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

131925-1L 131925-500ML
BT131925-1LEA 57 USD
BT131925-1L BT131925-500ML
Potassium hydroxide 30% w/v water
Potassium hydroxide

CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553; RTECS: TT2100000 UN No: UN1814; Haz Class: 8; Packing Group: II Liquid; Linear Formula: KOH; MW: 56.11 Density (g/mL): 1.29


  • RTECS: TT2100000
  • Liquid

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.29 g/mL
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR