Trichloroethylene ≥99.5% ACS

Supplier: BeanTown Chemical

131840-1L 131840-4L 131840-500ML
BT131840-1LEA 57.74 USD
BT131840-1L BT131840-4L BT131840-500ML
Trichloroethylene ≥99.5% ACS
Trichloroethylene

CAS: 79-01-6; EC No: 201-167-4; MDL No: MFCD00000838; RTECS: KX4550000 UN No: UN1710; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: ClCH=CCl2 ; Molecular Formula: C2HCl3; MW: 131.39 Melting Point: -85°; Boiling Point: 87° Density (g/mL): 1.463; Refractive Index: 1.476 Light Sensitive


  • RTECS: KX4550000
  • Refractive index: 1.476
  • Light Sensitive
  • Liquid

Formula: ClCH=CCl₂
Boiling Pt: 87 °C
Melting Pt: –85 °C
Density: 1.463 g/mL
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ac57d7a9ea2e3097408c3559993eb33587aec09&vid=767d7d09973e747c5a2d07a4d46dffedfb559ceb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% min.
Color (APHA) ≤10ppm
Residue after evaporation ≤10ppm
Titrable acid ≤0.0001meq/g
Titrable base ≤0.0003meq/g
Water ≤200ppm
Heavy metals ≤1ppm
Free halogens Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR