Boron (≥95% B-11), crystalline powder -100 mesh
Supplier: BeanTown Chemical
131725-25G
131725-5G
BT131725-25GEA
1614
USD
BT131725-25G
BT131725-5G
Boron (≥95% B-11), crystalline powder -100 mesh
Boron
CAS: 7440-42-8; EC No: 231-151-2; MDL No: MFCD00134034; RTECS: ED7350000
UN No: UN3178; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: B; MW: 10.81
Melting Point: 2076°; Boiling Point: 3927°
Density (g/mL): 2.34
- RTECS: ED7350000
- Powder
Formula:
B Boiling Pt: 3927 °C Melting Pt: 2076 °C Density: 2.34 g/mL |
MDL Number:
MFCD00134034 CAS Number: 7440-42-8 EINECS: 231-151-2 UN: 3178 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...