Carbon nanotube multi-walled as-produced cathode deposits
Supplier: BeanTown Chemical
131370-1G
131370-5G
BT131370-1GEA
55
USD
BT131370-1G
BT131370-5G
Carbon nanotube multi-walled as-produced cathode deposits
Carbon nanotube
CAS: 308068-56-6; MDL No: MFCD00133992
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: C; MW: 12.011
- Powder
Formula:
C |
MDL Number:
MFCD00133992 CAS Number: 308068-56-6 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...