Potassium hydroxide ≥85%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

131115-10KG 131115-2.5KG 131115-500G
BT131115-10KGEA 247 USD
BT131115-10KG BT131115-2.5KG BT131115-500G
Potassium hydroxide ≥85%, Technical Grade
Potassium hydroxide

CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553; RTECS: TT2100000 UN No: UN1813; Haz Class: 8; Packing Group: II Flakes; Linear Formula: KOH; MW: 56.11 Melting Point: 361°; Boiling Point: 1320° Density (g/mL): 2.044 Air Sensitive , Hygroscopic


  • RTECS: TT2100000
  • Air Sensitive, Hygroscopic
  • Flakes

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.044 g/mL
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Assay 85%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR