2,3,6-Trifluoroaniline ≥99%

Supplier: BeanTown Chemical

130720-1G 130720-5G
BT130720-1GEA 38 USD
BT130720-1G BT130720-5G
2,3,6-Trifluoroaniline ≥99%
2,3,6-Trifluoroaniline

CAS: 67815-56-9; MDL No: MFCD00012369 UN No: UN2941; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H4F3N; MW: 147.10 Flash point: 61°C (141°F) Density (g/mL): 1.39


  • Liquid

Formula: C₆H₄F₃N
MW: 147.1
Density: 1.39 g/mL
Flash Pt: 61 °C (141 °F)
MDL Number: MFCD00012369
CAS Number: 67815-56-9
UN: 2941
ADR: 6.1 III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR