Tetraethyl orthosilicate (TEOS) ≥99%
Supplier: BeanTown Chemical
Synonyms:
Tetraethoxysilane
130455-2.5KG
130455-500G
BT130455-2.5KGEA
326.55
USD
BT130455-2.5KG
BT130455-500G
Tetraethyl orthosilicate (TEOS) ≥99%
Tetraethyl orthosilicate (TEOS)
CAS: 78-10-4; EC No: 201-083-8; MDL No: MFCD00009062; RTECS: VV9450000
UN No: UN1292; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: Si(OCH2CH3)4; Molecular Formula: C8H20O4Si; MW: 208.33
Melting Point: -85°; Boiling Point: 168°; Flash point: 46°C (116°F)
Density (g/mL): 0.934; Refractive Index: 1.382
Moisture Sensitive
- RTECS: VV9450000
- Refractive index: 1.382
- Moisture Sensitive
- Liquid
Formula:
Si(OC₂H₅)₄ Boiling Pt: 168 °C Melting Pt: –85 °C Density: 0.934 g/mL Flash Pt: 46 °C (116 °F) |
MDL Number:
MFCD00009062 CAS Number: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...