Phenol ≥99.0% stabilised ACS
Supplier: BeanTown Chemical
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
130435-100G
130435-2KG
130435-500G
BT130435-100GEA
57.72
USD
BT130435-100G
BT130435-2KG
BT130435-500G
Phenol ≥99.0% stabilised ACS
Phenol
CAS: 108-95-2; EC No: 203-632-7; MDL No: MFCD00002143; RTECS: SJ3325000
UN No: UN1671; Haz Class: 6.1; Packing Group: II
Crystalline; Linear Formula: C6H5OH; MW: 94.11
Melting Point: 40-42°; Boiling Point: 182°; Flash point: 79°C (174°F)
Density (g/mL): 1.071
Air Sensitive, Hygroscopic, Light Sensitive
- RTECS: SJ3325000
- Air Sensitive, Hygroscopic, Light Sensitive
- Crystalline
Formula:
C₆H₅OH Boiling Pt: 182 °C Melting Pt: 40…42 °C Density: 1.071 g/mL Flash Pt: 79 °C (174 °F) |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1,II |
Specification Test Results
Assay | 99.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...