3-Pentanone ≥99%
Supplier: BeanTown Chemical
Synonyms:
Diethyl ketone
130240-100ML
130240-2.5L
130240-500ML
BT130240-100MLEA
42.91
USD
BT130240-100ML
BT130240-2.5L
BT130240-500ML
3-Pentanone ≥99%
3-Pentanone
CAS: 96-22-0; EC No: 202-490-3; MDL No: MFCD00009320; RTECS: SA8050000
UN No: UN1156; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2COCH2CH3; Molecular Formula: C5H10O; MW: 86.13
Melting Point: -42°; Boiling Point: 101°; Flash point: 7°C (45°F)
Density (g/mL): 0.813; Refractive Index: 1.392
- RTECS: SA8050000
- Refractive index: 1.392
- Liquid
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 101 Melting Pt: -40 Density: 0.813 g/mL Flash Pt: 7 °C (45 °F) |
MDL Number:
MFCD00009320 CAS Number: 96-22-0 EINECS: 202-490-3 UN: 1156 ADR: 3 II Merck Index: 13,03148 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...