Triethoxysilane ≥96%

Supplier: BeanTown Chemical

130060-10G 130060-250G 130060-50G
BT130060-10GEA 42.57 USD
BT130060-10G BT130060-250G BT130060-50G
Triethoxysilane ≥96%
Triethoxysilane

CAS: 998-30-1; EC No: 213-650-7; MDL No: MFCD00009061; RTECS: VV6682000 UN No: UN3384; Haz Class: 6.1 (3); Packing Group: I Liquid; Linear Formula: (CH3CH2O)3SiH; Molecular Formula: C6H16O3Si; MW: 164.28 Melting Point: -170°; Boiling Point: 134-135°; Flash point: 26°C (79°F) Density (g/mL): 0.885 ; Refractive Index: 1.377 Moisture Sensitive


  • RTECS: VV6682000
  • Refractive index: 1.377
  • Moisture Sensitive
  • Liquid

Formula: C₆H₁₆O₃Si
MW: 164.28 g/mol
Boiling Pt: 135
Melting Pt: -170
Density: 0.885 g/mL
Flash Pt: 26 °C (79 °F)
MDL Number: MFCD00009061
CAS Number: 998-30-1
EINECS: 213-650-7
UN: 3384
ADR: 6.1 I

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR