Tin(II) chloride dihydrate 98.0-103.0% ACS

Supplier: BeanTown Chemical

Synonyms: Stannous chloride dihydrate

130030-100G 130030-2KG 130030-500G
BT130030-100GEA 80.12 USD
BT130030-100G BT130030-2KG BT130030-500G
Tin(II) chloride dihydrate 98.0-103.0% ACS
Tin (II) chloride dihydrate

CAS: 10025-69-1; EC No: 231-868-0; MDL No: MFCD00149863; RTECS: XP8850000 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: SnCl2·2H2O; MW: 225.65 Melting Point: 37-38° (decomposes); Boiling Point: 652° Density (g/mL): 2.71 Air Sensitive, Moisture Sensitive


  • RTECS: XP8850000
  • Air Sensitive, Moisture Sensitive
  • Crystalline

Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623
Melting Pt: 246 (dec.)
Density: 2.71 g/mL
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay 98.0-103.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR