Instrument Calibration(ICAL) Standards Kit

Supplier: BeanTown Chemical
129930-100ML
BT129930-100MLEA 287.79 USD
BT129930-100ML
Instrument Calibration(ICAL) Standards Kit
Standards Cationic Standards Multi-Element Standards
MDL No: MFCD00147264

UN No: UN3264; Haz Class: 8; Packing Group: II
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR