Barium nitrate ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

129835-1G 129835-25G 129835-5G
BT129835-1GEA 53 USD
BT129835-1G BT129835-25G BT129835-5G
Barium nitrate ≥99.999% (trace metals basis)
Barium nitrate

CAS: 10022-31-8; EC No: 233-020-5; MDL No: MFCD00003442; RTECS: CQ9625000 UN No: UN1446; Haz Class: 5.1 (6.1); Packing Group: II Crystalline; Linear Formula: Ba(NO3)2 ; MW: 261.34 Melting Point: 592° (decomposes) Hygroscopic


  • RTECS: CQ9625000
  • Hygroscopic
  • Crystalline

Formula: Ba(NO₃)₂
Melting Pt: 592 °C (dec.)
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR