Iodoethane ≥98% stabilised
Supplier: BeanTown Chemical
Synonyms:
Ethyl iodide
129705-100G
129705-2.5KG
129705-500G
BT129705-100GEA
44.36
USD
BT129705-100G
BT129705-2.5KG
BT129705-500G
Iodoethane ≥98% stabilised
Iodoethane
CAS: 75-03-6; EC No: 200-833-1; MDL No: MFCD00001091; RTECS: KI4750000
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: CH3CH2I; MW: 155.97
Melting Point: -108°; Boiling Point: 69-73°; Flash point: 53°C (127°F)
Density (g/mL): 1.94; Refractive Index: 1.513
Light Sensitive
- RTECS: KI4750000
- Refractive index: 1.513
- Light Sensitive
- Liquid
Formula:
C₂H₅I MW: 155.97 g/mol Boiling Pt: 72 Melting Pt: -108 Density: 1.94 g/mL Flash Pt: 53 °C (127 °F) |
MDL Number:
MFCD00001091 CAS Number: 75-03-6 EINECS: 200-833-1 UN: 2810 ADR: 6.1 III Merck Index: 13,03846 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...