Potassium cyanide ≥96.0% ACS
Supplier: BeanTown Chemical
128615-1KG
128615-250G
128615-50G
BT128615-1KGEA
176.32
USD
BT128615-1KG
BT128615-250G
BT128615-50G
Potassium cyanide ≥96.0% ACS
Potassium cyanide
CAS: 151-50-8; EC No: 205-792-3; MDL No: MFCD00011397; RTECS: TS8750000
UN No: UN1680; Haz Class: 6.1; Packing Group: I
Solid; Linear Formula: KCN; MW: 65.12
Melting Point: 634°; Boiling Point: 1625°
Density (g/mL): 1.52
- RTECS: TS8750000
- Solid
Formula:
KCN Boiling Pt: 1625 °C Melting Pt: 634 °C Density: 1.52 g/mL |
MDL Number:
MFCD00011397 CAS Number: 151-50-8 EINECS: 205-792-3 UN: 1680 ADR: 6.1,I |
Specification Test Results
Assay | 96.0% min. |
Chloride (Cl) | ≤0.5% |
Phosphate (PO4) | ≤0.005% |
Sulfate (SO4) | ≤0.04% |
Sulfide (S) | ≤0.003% |
Thiocyanate (SCN) | Passes test |
Iron (Fe) | ≤0.03% |
Lead (Pb) | ≤2ppm |
Sodium (Na) | ≤0.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...