Acetone ≥99.5% for HPLC

Supplier: BeanTown Chemical

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

128560-1L 128560-4L 128560-4X1L 128560-4X4L
BT128560-1LEA 50.18 USD
BT128560-1L BT128560-4L BT128560-4X1L BT128560-4X4L
Acetone ≥99.5% for HPLC
Acetone

CAS: 67-64-1; EC No: 200-662-2; MDL No: MFCD00008765; RTECS: AL3150000 UN No: UN1090; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3COCH3; MW: 58.08 Melting Point: -94°; Boiling Point: 56°; Flash point: -17°C (1°F) Density (g/mL): 0.791; Refractive Index: 1.359


  • RTECS: AL3150000
  • Refractive index: 1.359
  • Liquid

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.791 g/mL
Flash Pt: –17 °C (1 °F)
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR