Diethylzinc 15% (w/w) in hexane
Supplier: BeanTown Chemical
128505-100G
128505-25G
128505-500G
BT128505-100GEA
129.46
USD
BT128505-100G
BT128505-25G
BT128505-500G
Diethylzinc 15% (w/w) in hexane
Diethylzinc
CAS: 557-20-0; EC No: 209-161-3; MDL No: MFCD00009021
UN No: UN3394; Haz Class: 4.2 (4.3); Packing Group: I
Liquid; Linear Formula: (CH3CH2)2Zn; MW: 123.49
Flash point: -40°C (-40°F)
Density (g/mL): 0.720
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₄H₁₀Zn Density: 0.720 g/mL Flash Pt: –40 °C (–40 °F) |
MDL Number:
MFCD00009021 CAS Number: 557-20-0 EINECS: 209-161-3 ADR: 4.2 (4.3),I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...