Aluminium ≥99.7%, soluble APS 11 µm

Supplier: BeanTown Chemical

Synonyms: Aluminum

128475-1KG 128475-250G 128475-5KG
BT128475-1KGEA 152 USD
BT128475-1KG BT128475-250G BT128475-5KG
Aluminium ≥99.7%, soluble APS 11 µm
Aluminium

CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330000 UN No: UN3208; Haz Class: 4.3; Packing Group: II Solid; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7


  • RTECS: BD0330000
  • Solid

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
ADR: 4.3 II
Merck Index: 13,00321

Order Now

Specification Test Results

Assay 99.7%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR