Propionaldehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: Propanal

128270-1L 128270-500ML 128270-5L
BT128270-1LEA 41.42 USD
BT128270-1L BT128270-500ML BT128270-5L
Propionaldehyde ≥97%
Propionaldehyde

CAS: 123-38-6; EC No: 204-623-0; MDL No: MFCD00007020; RTECS: UE0350000 UN No: UN1275; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH2CHO; Molecular Formula: C3H6O; MW: 58.08 Melting Point: -81°; Boiling Point: 46-50°; Flash point: -40°C (-40°F) Density (g/mL): 0.805; Refractive Index: 1.362 Air Sensitive


  • RTECS: UE0350000
  • Refractive index: 1.362
  • Air Sensitive
  • Liquid

Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48
Melting Pt: -81
Density: 0.805 g/mL
Flash Pt: –40 °C (–40 °F)
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3 II
Merck Index: 13,07915

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR