Lithium nitrate, anhydrous ≥99%

Supplier: BeanTown Chemical

128175-1KG 128175-250G 128175-25G
BT128175-1KGEA 327.15 USD
BT128175-1KG BT128175-250G BT128175-25G
Lithium nitrate, anhydrous ≥99%
Lithium nitrate

CAS: 7790-69-4; EC No: 232-218-9; MDL No: MFCD00011094 UN No: UN2722; Haz Class: 5.1; Packing Group: III Crystalline; Molecular Formula: LiNO3 ; MW: 68.95 Melting Point: 264° Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: LiNO₃
Melting Pt: 264 °C
MDL Number: MFCD00011094
CAS Number: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR