n-Heptane ≥96.0% environmental
Supplier: BeanTown Chemical
CAS: 142-82-5; EC No: 205-563-8; MDL No: MFCD00009544; RTECS: MI7700000
UN No: UN1206; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3(CH2)5CH3; Molecular Formula: C7H16 ; MW: 100.21
Melting Point: -91°; Boiling Point: 97-99°; Flash point: -4°C (24°F)
Density (g/mL): 0.684; Refractive Index: 1.387
- RTECS: MI7700000
- Refractive index: 1.387
- Liquid
Formula:
H₃C(CH₂)₅CH₃ Boiling Pt: 97…99 °C Melting Pt: –91 °C Density: 0.684 g/mL Flash Pt: –4 °C (24 °F) |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3,II |
|
Specification Test Results
Assay | 96.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...