Phosphorous acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
Phosphonic acid
127760-2KG
127760-500G
BT127760-2KGEA
239.72
USD
BT127760-2KG
BT127760-500G
Phosphorous acid ≥97%
Phosphorous acid
CAS: 13598-36-2; EC No: 237-066-7; MDL No: MFCD00137258; RTECS: SZ6400000
UN No: UN2834; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: H3PO3; MW: 82.00
Melting Point: 74°; Boiling Point: 200° (decomposes)
Density (g/mL): 1.651
Air Sensitive , Hygroscopic
- RTECS: SZ6400000
- Air Sensitive, Hygroscopic
- Crystalline
Formula:
H₃O₃P MW: 82 g/mol Boiling Pt: 200 °C (dec.) Melting Pt: 74 °C Density: 1.651 g/mL |
MDL Number:
MFCD00137258 CAS Number: 13598-36-2 EINECS: 237-066-7 UN: 2834 ADR: 8 III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...