Cobalt ≥99.995% (trace metals basis), powder -22 mesh

Supplier: BeanTown Chemical

127635-25G 127635-5G
BT127635-25GEA 281 USD
BT127635-25G BT127635-5G
Cobalt ≥99.995% (trace metals basis), powder -22 mesh
Cobalt

CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GF8750000 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Co; MW: 58.93 Melting Point: 1495°; Boiling Point: 2900° Density (g/mL): 8.92


  • RTECS: GF8750000
  • Powder

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.92 g/mL
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1 III
Merck Index: 12,02488

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR