Decacarbonyldirhenium ≥96%

Supplier: BeanTown Chemical

Synonyms: Rhenium carbonyl

127420-1G 127420-250MG
BT127420-1GEA 102 USD
BT127420-1G BT127420-250MG
Decacarbonyldirhenium ≥96%
Decacarbonyldirhenium

CAS: 14285-68-8; EC No: 238-202-8; MDL No: MFCD00011198 UN No: UN3466; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: Re2(CO)10; MW: 652.52 Melting Point: 170° (decomposes)


  • Powder

Formula: C₁₀O₁₀Re₂
MW: 652.52
Melting Pt: 170 °C (dec.)
Density: 2.87
MDL Number: MFCD00011198
CAS Number: 14285-68-8
EINECS: 238-202-8
UN: 3466
ADR: 6.1 III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR