Platinum(IV) chloride ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Platinum tetrachloride

127405-1G 127405-5G
BT127405-1GEA 189 USD
BT127405-1G BT127405-5G
Platinum(IV) chloride ≥99.99%
Platinum (IV) chloride

CAS: 13454-96-1; EC No: 236-645-1; MDL No: MFCD00011182 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline/Powder; Linear Formula: PtCl4; MW: 336.89 Melting Point: 370° (decomposes) Density (g/mL): 4.303 Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: Cl₄Pt
MW: 336.89 g/mol
Melting Pt: 370 (dec.)
Density: 4.303 g/mL
MDL Number: MFCD00011182
CAS Number: 13454-96-1
EINECS: 236-645-1
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR