Platinum, black, high surface area

Supplier: BeanTown Chemical

Synonyms: Platinium

127255-1G 127255-250MG 127255-5G
BT127255-1GEA 311 USD
BT127255-1G BT127255-250MG BT127255-5G
Platinum, black, high surface area
Platinum

CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000 UN No: UN3089; Haz Class: 4.1; Packing Group: II Solid, S.A. 25.0-30.4 m2/g ; Molecular Formula: Pt; MW: 195.08 Melting Point: 1772°; Boiling Point: 3827° Density (g/mL): 21.45


  • RTECS: TP2160000
  • Solid, S.A. 25.0-30.4 m²/g

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45 g/mL
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
ADR: 4.1 II
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR