Nickel(II) chloride, anhydrous ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Nickel dichloride

127075-25G 127075-5G
BT127075-25GEA 478.23 USD
BT127075-25G BT127075-5G
Nickel(II) chloride, anhydrous ≥99.99%
Nickel (II) chloride

CAS: 7718-54-9; EC No: 231-743-0; MDL No: MFCD00011142; RTECS: QR6475000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: NiCl2; MW: 129.60 Melting Point: 1001° Density (g/mL): 3.55 Hygroscopic


  • RTECS: QR6475000
  • Hygroscopic
  • Crystalline/Powder

Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1009
Density: 3.55 g/mL
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 12,06588

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR