Toluene, anhydrous ≥99.5%
Supplier: BeanTown Chemical
CAS: 108-88-3; EC No: 203-625-9; MDL No: MFCD00008512; RTECS: XS5250000
UN No: UN1294; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: C6H5CH3; Molecular Formula: C7H8; MW: 92.14
Melting Point: -93°; Boiling Point: 110-111°; Flash point: 4°C (39°F)
Density (g/mL): 0.865; Refractive Index: 1.496
- RTECS: XS5250000
- Refractive index: 1.496
- Liquid
Formula:
C₆H₅CH₃ Boiling Pt: 110…111 °C Melting Pt: –93 °C Density: 0.865 g/mL Flash Pt: 4 °C (39 °F) |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II |
|
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...