Silver ≥99.9%, soluble 80% <20 µm

Supplier: BeanTown Chemical

126845-10G 126845-250G 126845-50G
BT126845-10GEA 66.82 USD
BT126845-10G BT126845-250G BT126845-50G
Silver ≥99.9%, soluble 80% <20 µm
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397 UN No: UN3077; Haz Class: 9; Packing Group: III Solid, Scott density 18-28 g/in3, S.A. 1.0-2.2 m2/g; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • Solid, Scott density 18-28 g/in3, S.A. 1.0-2.2 m²/g

Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960 °C
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
ADR: 9,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR