tetra-Chloroauric(III) acid hydrate ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

126735-1G 126735-5G
BT126735-1GEA 152.97 USD
BT126735-1G BT126735-5G
tetra-Chloroauric(III) acid hydrate ≥99.9%
tetra-Chloroauric (III) acid hydrate

CAS: 27988-77-8; EC No: 240-948-4; MDL No: MFCD00149903; RTECS: MD5428000 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Molecular Formula: HAuCl4·xH2O; MW: 339.79 (anhydrous) Hygroscopic , Light Sensitive


  • RTECS: MD5428000
  • Hygroscopic, Light Sensitive
  • Crystalline

Formula: HAuCl₄·H₂O
MW: 357.8 g/mol
MDL Number: MFCD00149903
CAS Number: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR