Bismuth(III) fluoride ≥99.999%
Supplier: BeanTown Chemical
126545-100G
126545-25G
BT126545-100GEA
801
USD
BT126545-100G
BT126545-25G
Bismuth(III) fluoride ≥99.999%
Bismuth (III) fluoride
CAS: 7787-61-3; EC No: 232-124-8; MDL No: MFCD00010893
UN No: UN3260; Haz Class: 8; Packing Group: III
-200 Mesh Powder; Molecular Formula: BiF3; MW: 265.98
Hygroscopic
- Hygroscopic
- -200 Mesh Powder
Formula:
BiF₃ MW: 265.98 Boiling Pt: 230 Density: 3.7 |
MDL Number:
MFCD00010893 CAS Number: 7787-61-3 EINECS: 232-124-8 UN: 3260 ADR: 8 III Merck Index: 12,01305 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...