Ytterbium ≥99.9% (trace rare earth metals basis), pieces, sublimated

Supplier: BeanTown Chemical

126360-100G 126360-25G 126360-5G
BT126360-100GEA 936.13 USD
BT126360-100G BT126360-25G BT126360-5G
Ytterbium ≥99.9% (trace rare earth metals basis), pieces, sublimated
Ytterbium

CAS: 7440-64-4; EC No: 231-173-2; MDL No: MFCD00011286; RTECS: ZG1925000 UN No: UN3208; Haz Class: 4.3; Packing Group: II Solid, packaged under argon; Molecular Formula: Yb; MW: 173.04 Melting Point: 824°; Boiling Point: 1196° Density (g/mL): 6.54


  • RTECS: ZG1925000
  • Solid, packaged under argon

Formula: Yb
MW: 173.04 g/mol
Boiling Pt: 1427
Melting Pt: 824
Density: 6.54 g/mL
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
ADR: 4.3 II
Merck Index: 12,10239

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR