Cobalt(II) oxide ≥95%

Supplier: BeanTown Chemical

Synonyms: Cobalt oxide

126335-100G 126335-25G
BT126335-100GEA 276.55 USD
BT126335-100G BT126335-25G
Cobalt(II) oxide ≥95%
Cobalt (II) oxide

CAS: 1307-96-6; EC No: 215-154-6; MDL No: MFCD00016031; RTECS: GG2800000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: CoO; MW: 74.93 Melting Point: 1935° Air Sensitive


  • RTECS: GG2800000
  • Air Sensitive
  • Powder

Formula: CoO
MW: 74.93 g/mol
Melting Pt: 1933
MDL Number: MFCD00016031
CAS Number: 1307-96-6
EINECS: 215-154-6
UN: 2811
ADR: 6.1 III
Merck Index: 12,02507

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR