(Chloromethylene)dimethylammonium chloride ≥96%

Supplier: BeanTown Chemical

Synonyms: N-Chloromethylen-N,N-Dimethyl Ammonium Chloride

124570-25G 124570-5G
BT124570-25GEA 158.63 USD
BT124570-25G BT124570-5G
(Chloromethylene)dimethylammonium chloride ≥96%
(Chloromethylene)dimethylammonium chloride

CAS: 3724-43-4; EC No: 425-970-6; MDL No: MFCD00011868 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline/Powder; Linear Formula: ClCH=N(CH3)2Cl; Molecular Formula: C3H7Cl2N; MW: 128.00 Melting Point: 132° (decomposes) Moisture Sensitive


  • Moisture Sensitive
  • Crystalline/Powder

Formula: C₃H₇Cl₂N
MW: 128.00 g/mol
Melting Pt: 132 °C (dec.)
MDL Number: MFCD00011868
CAS Number: 3724-43-4
EINECS: 425-970-6
ADR: 8 II

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR