Cyclohexane ≥99% for HPLC

Supplier: BeanTown Chemical

124330-4L 124330-4X1L
BT124330-1LEA 57 USD
BT124330-1L BT124330-4L BT124330-4X1L
Cyclohexane ≥99% for HPLC
Cyclohexane

CAS: 110-82-7; EC No: 203-806-2; MDL No: MFCD00003814; RTECS: GU6300000 UN No: UN1145; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C6H12; MW: 84.16 Melting Point: 4-7°; Boiling Point: 80.7°; Flash point: -18°C (-1°F) Density (g/mL): 0.779; Refractive Index: 1.426


  • RTECS: GU6300000
  • Refractive index: 1.426
  • Liquid

Formula: C₆H₁₂
Boiling Pt: 80.7 °C
Melting Pt: 4…7 °C
Density: 0.779 g/mL
Flash Pt: –18 °C (–1 °F)
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR