Ruthenium ≥99.99% (trace metals basis excluding Ca), powder -200 mesh

Supplier: BeanTown Chemical

124135-1G 124135-25G 124135-5G
BT124135-1GEA 155 USD
BT124135-1G BT124135-25G BT124135-5G
Ruthenium ≥99.99% (trace metals basis excluding Ca), powder -200 mesh
Ruthenium

CAS: 7440-18-8; EC No: 231-127-1; MDL No: MFCD00011207 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Ru; MW: 101.07 Melting Point: 2310°; Boiling Point: 3900° Density (g/mL): 12.45


  • Powder

Formula: Ru
Boiling Pt: 3900 °C
Melting Pt: 2310 °C
Density: 12.45 g/mL
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR