Nitrobenzene ≥99%
Supplier: BeanTown Chemical
CAS: 98-95-3; EC No: 202-716-0; MDL No: MFCD00007043; RTECS: DA6475000
UN No: UN1662; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C6H5NO2; MW: 123.11
Melting Point: 5-6°; Boiling Point: 210-211°; Flash point: 87°C (188°F)
Density (g/mL): 1.196; Refractive Index: 1.551
- RTECS: DA6475000
- Refractive index: 1.551
- Liquid
Formula:
C₆H₅NO₂ Boiling Pt: 210…211 °C Melting Pt: 5…6 °C Density: 1.196 g/mL Flash Pt: 87 °C (188 °F) |
MDL Number:
MFCD00007043 CAS Number: 98-95-3 EINECS: 202-716-0 UN: 1662 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...