Potassium tetrachloroplatinate(II) ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate

123400-1G 123400-5G
BT123400-1GEA 136 USD
BT123400-1G BT123400-5G
Potassium tetrachloroplatinate(II) ≥99.9%
Potassium tetrachloroplatinate (II)

CAS: 10025-99-7; EC No: 233-050-9; MDL No: MFCD00011378; RTECS: TP1850000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: K2PtCl4; MW: 415.09 Density (g/mL): 3.38


  • RTECS: TP1850000
  • Crystalline/Powder

Formula: Cl₄K₂Pt
MW: 415.09
Melting Pt: 265
Density: 3.38 g/mL
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1 III
Merck Index: 12,07855

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR