Silver(I) oxide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
di-Silver oxide, Disilver oxide, Silver oxide
123325-1G
123325-25G
123325-5G
BT123325-1GEA
23
USD
BT123325-1G
BT123325-25G
BT123325-5G
Silver(I) oxide ≥99.99%
Silver(I) oxide
CAS: 20667-12-3; EC No: 243-957-1; MDL No: MFCD00003404; RTECS: VW4900000
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Powder; Molecular Formula: Ag2O; MW: 231.74
Melting Point: 230° (decomposes)
Density (g/mL): 7.143
Light Sensitive
- RTECS: VW4900000
- Powder
- Light Sensitive
Formula:
Ag₂O Melting Pt: 230 °C (dec.) Density: 7.143 g/mL |
MDL Number:
MFCD00003404 CAS Number: 20667-12-3 UN: 1479 ADR: 5.1,II |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...